Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01015880_m1
          See other SETD2 GE Assays ›
          Gene Symbol:
          SETD2
          Gene Name:
          SET domain containing 2
          Gene Aliases:
          HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP
          Chromosome Location:
          Chr.3: 47016408 - 47164109 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01015880_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SETD2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014159.6 NP_054878.5 13 - 14 6160 100
          XM_011533631.2 XP_011531933.1 14 - 15 7656 100
          XM_011533632.2 XP_011531934.1 13 - 14 6232 100
          XM_011533633.2 XP_011531935.1 13 - 14 7523 100
          XM_011533634.1 XP_011531936.1 13 - 14 6185 100
          XM_017006270.1 XP_016861759.1 11 - 12 7359 100
          GenBank mRNA AB051519.2 - 11 - 12 4160 100
          AJ238403.1 - 11 - 12 5007 100
          AY576987.1 - 11 - 12 4669 100
          BC090954.1 - 11 - 12 3950 100
          BC117162.1 - 11 - 12 4783 40125713 100
          BC117164.1 - 11 - 12 4783 40125715 100
          BX649110.1 - 11 - 12 5927 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010968.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001364.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SETD2

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          morphogenesis of a branching structure
          neural tube closure
          mismatch repair
          regulation of transcription, DNA-templated
          transcription elongation from RNA polymerase II promoter
          regulation of mRNA export from nucleus
          forebrain development
          nucleosome organization
          cell migration involved in vasculogenesis
          mesoderm morphogenesis
          embryonic cranial skeleton morphogenesis
          stem cell development
          pericardium development
          embryonic placenta morphogenesis
          coronary vasculature morphogenesis
          histone H3-K36 trimethylation
          histone H3-K36 dimethylation
          protein binding
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K36 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03023385.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023385.hg.1 ENST00000451092.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023385.hg.1 ENST00000543224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023385.hg.1 NM_014159.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03023385.hg.1 TC03001364.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 ENST00000451092.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 ENST00000492397.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 ENST00000543224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 NM_014159.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03023387.hg.1 TC03001364.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000330022.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000431180.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000445387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 NM_014159.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 TC0300010968.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000445387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000330022.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 ENST00000431180.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180069.hg.1 NM_014159.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000330022.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000431180.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000445387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000492397.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 NM_014159.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 TC0300010968.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000330022.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000409792.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000431180.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000445387.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 ENST00000492397.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180073.hg.1 NM_014159.6 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0