Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01017875_m1
          See other PDK3 GE Assays ›
          Gene Symbol:
          PDK3
          Gene Name:
          pyruvate dehydrogenase kinase 3
          Gene Aliases:
          CMTX6, GS1-358P8.4
          Chromosome Location:
          Chr.X: 24465227 - 24550466 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01017875_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDK3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142386.2 NP_001135858.1 7 - 8 981 92
          NM_005391.4 NP_005382.1 7 - 8 981 92
          GenBank mRNA AK301965.1 - 7 - 8 984 92
          BC015948.1 - 7 - 8 928 3922929 92
          L42452.1 - 7 - 8 774 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00006816.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000118.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDK3

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          regulation of acetyl-CoA biosynthetic process from pyruvate
          regulation of glucose metabolic process
          peptidyl-serine phosphorylation
          peroxisome proliferator activated receptor signaling pathway
          cellular response to glucose stimulus
          cellular response to fatty acid
          hypoxia-inducible factor-1alpha signaling pathway
          regulation of reactive oxygen species metabolic process
          protein kinase activity
          protein serine/threonine kinase activity
          pyruvate dehydrogenase (acetyl-transferring) kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X001589.hg.1 ENST00000379162.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001589.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001589.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001589.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001589.hg.1 TC0X000118.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 ENST00000568479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001588.hg.1 ENST00000379162.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001588.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X001588.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001588.hg.1 NM_005391.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X001588.hg.1 TC0X000118.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 TC0X00006816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146685.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 TC0X00006816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00146684.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000379162.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000441463.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000493226.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 ENST00000568479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_001142386.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00006816.hg.1 NM_005391.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix