Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01019709_g1
          See other MYO15B GE Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01019709_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001309242.1 NP_001296171.1
          51 - 52 7895 65
          XM_017025120.1 XP_016880609.1
          53 - 54 7602 65
          XM_017025121.1 XP_016880610.1
          53 - 54 7587 65
          XM_017025122.1 XP_016880611.1
          53 - 54 7566 65
          XM_017025123.1 XP_016880612.1
          53 - 54 7604 65
          XM_017025124.1 XP_016880613.1
          52 - 53 7476 65
          XM_017025125.1 XP_016880614.1
          51 - 52 7325 65
          XM_017025126.1 XP_016880615.1
          48 - 49 7067 65
          XM_017025127.1 XP_016880616.1
          51 - 52 6910 65
          XM_017025128.1 XP_016880617.1
          52 - 53 6656 65
          XM_017025129.1 XP_016880618.1
          51 - 52 5888 65
          XM_017025130.1 XP_016880619.1
          50 - 51 5844 65
          XM_017025131.1 XP_016880620.1
          49 - 50 6283 65
          XM_017025132.1 XP_016880621.1
          48 - 49 6224 65
          XM_017025133.1 XP_016880622.1
          46 - 47 5749 65
          XM_017025134.1 XP_016880623.1
          47 - 48 6110 65
          XM_017025135.1 XP_016880624.1
          41 - 42 4766 65
          XM_017025136.1 XP_016880625.1
          41 - 42 5115 65
          XM_017025137.1 XP_016880626.1
          38 - 39 4660 65
          XM_017025138.1 XP_016880627.1
          33 - 34 4156 65
          XM_017025139.1 XP_016880628.1
          34 - 35 3957 65
          XM_017025140.1 XP_016880629.1
          31 - 32 3770 65
          XM_017025143.1 XP_016880632.1
          26 - 27 3047 65
          XM_017025144.1 XP_016880633.1
          47 - 48 7530 65
          GenBank mRNA AF418292.1 -
          26 - 27 3505 65
          AF418293.1 -
          27 - 28 3111 65
          AK026339.1 -
          3 - 4 210 65
          AL713730.1 -
          2 - 3 104 65
          BC027875.1 - LIT3316
          MYO15BP
          3 - 4 599 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CV029543.1 - 3 - 4 182 Chr. 17 - 75621133 - 75624449 [+] on Build GRCh38 65
          GenBankmRNA DA179389.1 - 3 - 4 417 Chr. 17 - 75620898 - 75623843 [+] on Build GRCh38 65
          GenBankmRNA DA382913.1 - 3 - 4 410 Chr. 17 - 75620905 - 75624020 [+] on Build GRCh38 65
          GenBankmRNA DA179408.1 - 3 - 4 417 Chr. 17 - 75620898 - 75623851 [+] on Build GRCh38 65
          GenBankmRNA CB155174.1 - 3 - 4 297 Chr. 17 - 75621018 - 75624050 [+] on Build GRCh38 65
          GenBankmRNA BM151468.1 - 2 - 3 Chr. 17 - 75621121 - 75623855 [-] on Build GRCh38 65
          GenBankmRNA BI772324.1 - 3 - 4 595 Chr. 17 - 75620722 - 75624006 [+] on Build GRCh38 65
          GenBankmRNA DA947560.1 - 4 - 5 465 Chr. 17 - 75620463 - 75623814 [+] on Build GRCh38 65
          GenBankmRNA DA932891.1 - 3 - 4 228 Chr. 17 - 75621087 - 75624250 [+] on Build GRCh38 65
          GenBankmRNA BM150946.1 - 2 - 3 Chr. 17 - 75621000 - 75623855 [-] on Build GRCh38 65
          GenBankmRNA DA634462.1 - 3 - 4 254 Chr. 17 - 75621061 - 75624214 [+] on Build GRCh38 65
          GenBankmRNA DA386191.1 - 3 - 4 443 Chr. 17 - 75620872 - 75623832 [+] on Build GRCh38 65
          GenBankmRNA BE247119.1 - 2 - 3 Chr. 17 - 75621074 - 75623855 [-] on Build GRCh38 65
          GenBankmRNA BM720779.1 - 2 - 3 105 Chr. 17 - 75621379 - 75624849 [+] on Build GRCh38 65
          GenBankmRNA DA058279.1 - 3 - 4 408 Chr. 17 - 75620907 - 75623964 [+] on Build GRCh38 65
          GenBankmRNA DA065143.1 - 2 - 3 513 Chr. 17 - 75620894 - 75622058 [+] on Build GRCh38 65
          GenBankmRNA BG912704.1 - 3 - 4 240 Chr. 17 - 75621075 - 75624899 [+] on Build GRCh38 65
          GenBankmRNA DA956318.1 - 3 - 4 420 Chr. 17 - 75620895 - 75623850 [+] on Build GRCh38 65
          GenBankmRNA BE245205.1 - 2 - 3 Chr. 17 - 75621345 - 75623855 [-] on Build GRCh38 65
          GenBankmRNA DN999110.1 - 3 - 4 297 Chr. 17 - 75621024 - 75624251 [+] on Build GRCh38 65
          GenBankmRNA BM150833.1 - 2 - 3 Chr. 17 - 75621032 - 75623855 [-] on Build GRCh38 65
          GenBankmRNA DA346664.1 - 3 - 4 420 Chr. 17 - 75620896 - 75623839 [+] on Build GRCh38 65
          GenBankmRNA DN997131.1 - 3 - 4 330 Chr. 17 - 75620992 - 75624269 [+] on Build GRCh38 65
          GenBankmRNA DB121220.1 - 3 - 4 299 Chr. 17 - 75621016 - 75624053 [+] on Build GRCh38 65
          GenBankmRNA DA209388.1 - 4 - 5 310 Chr. 17 - 75620332 - 75624058 [+] on Build GRCh38 65
          GenBankmRNA DB255318.1 - 3 - 4 419 Chr. 17 - 75620896 - 75623832 [+] on Build GRCh38 65
          GenBankmRNA DA958794.1 - 3 - 4 296 Chr. 17 - 75621019 - 75624064 [+] on Build GRCh38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LLGL2,
          scribble cell polarity complex component
          LLGL2 HGL,
          Hugl-2,
          LGL2
          -46.3kb Sense Chr. 17 - 75525080 - 75575209 on NCBI Build GRCh38
          Downstream RecQ like helicase 5 RECQL5 RECQ5 4.8kb Anti-sense Chr. 17 - 75626845 - 75667202 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008912.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYO15B

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000577613.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000577986.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000578220.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000580096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000610510.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000612587.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000621743.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000633867.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 NM_001309242.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 TC1700008912.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000577613.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000577986.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000578220.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000580096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000610510.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000612587.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000621743.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 ENST00000633867.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166964.hg.1 NM_001309242.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0