Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01019738_m1
          See other MYO15B GE Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01019738_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001309242.1 NP_001296171.1 42 - 43 6870 76
          XM_017025120.1 XP_016880609.1 44 - 45 6577 76
          XM_017025121.1 XP_016880610.1 44 - 45 6562 76
          XM_017025122.1 XP_016880611.1 44 - 45 6541 76
          XM_017025123.1 XP_016880612.1 44 - 45 6579 76
          XM_017025124.1 XP_016880613.1 43 - 44 6451 76
          XM_017025125.1 XP_016880614.1 42 - 43 6300 76
          XM_017025126.1 XP_016880615.1 39 - 40 6042 76
          XM_017025127.1 XP_016880616.1 42 - 43 5885 76
          XM_017025128.1 XP_016880617.1 43 - 44 5631 76
          XM_017025129.1 XP_016880618.1 42 - 43 4863 76
          XM_017025130.1 XP_016880619.1 41 - 42 4819 76
          XM_017025131.1 XP_016880620.1 40 - 41 5258 76
          XM_017025132.1 XP_016880621.1 39 - 40 5199 76
          XM_017025133.1 XP_016880622.1 37 - 38 4724 76
          XM_017025134.1 XP_016880623.1 38 - 39 5085 76
          XM_017025135.1 XP_016880624.1 32 - 33 3741 76
          XM_017025136.1 XP_016880625.1 32 - 33 4090 76
          XM_017025137.1 XP_016880626.1 29 - 30 3635 76
          XM_017025138.1 XP_016880627.1 24 - 25 3131 76
          XM_017025139.1 XP_016880628.1 25 - 26 2932 76
          XM_017025140.1 XP_016880629.1 22 - 23 2745 76
          XM_017025143.1 XP_016880632.1 17 - 18 2022 76
          XM_017025144.1 XP_016880633.1 38 - 39 6505 76
          GenBank mRNA AB058686.1 - 32 - 33 4977 76
          AF418286.1 - 38 - 39 6887 76
          AF418290.1 - 37 - 38 7297 76
          AK025259.1 - 1038 76
          AK097896.1 - 10 - 11 1346 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CR984271.1 - 2 - 3 156 Chr. 17 - 75617824 - 75620358 [+] on Build GRCh38 76
          GenBankmRNA DB138051.1 - 3 - 4 205 Chr. 17 - 75617270 - 75619805 [+] on Build GRCh38 76
          GenBankmRNA EL954657.1 - 4 - 5 460 Chr. 17 - 75616913 - 75619386 [+] on Build GRCh38 76
          GenBankmRNA DA316903.1 - 3 - 4 350 Chr. 17 - 75617125 - 75619454 [+] on Build GRCh38 76
          GenBankmRNA BI766913.1 - 522 Chr. - - [-] on Build 76
          GenBankmRNA CR986155.1 - 3 - 4 308 Chr. 17 - 75617167 - 75619796 [+] on Build GRCh38 76
          GenBankmRNA DA304564.1 - 2 - 3 166 Chr. 17 - 75617814 - 75619957 [+] on Build GRCh38 76
          GenBankmRNA DN831532.1 - Chr. - - [-] on Build 76
          GenBankmRNA DB134005.1 - 2 - 3 137 Chr. 17 - 75617843 - 75619964 [+] on Build GRCh38 76
          GenBankmRNA DA109947.1 - 2 - 3 501 Chr. 17 - 75617496 - 75619182 [+] on Build GRCh38 76
          GenBankmRNA DA428235.1 - 3 - 4 280 Chr. 17 - 75617580 - 75619565 [+] on Build GRCh38 76
          GenBankmRNA BI772088.1 - 423 Chr. - - [-] on Build 76
          GenBankmRNA DB138105.1 - 3 - 4 205 Chr. 17 - 75617270 - 75619805 [+] on Build GRCh38 76
          GenBankmRNA DA126806.1 - 5 - 6 552 Chr. 17 - 75616712 - 75619177 [+] on Build GRCh38 76
          GenBankmRNA BY798869.2 - 2 - 3 174 Chr. 17 - 75617810 - 75619955 [+] on Build GRCh38 76
          GenBankmRNA BI769659.1 - 5 - 6 562 Chr. 17 - 75616699 - 75619377 [+] on Build GRCh38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LLGL2,
          scribble cell polarity complex component
          LLGL2 HGL,
          Hugl-2,
          LGL2
          -42.9kb Sense Chr. 17 - 75525080 - 75575209 on NCBI Build GRCh38
          Downstream RecQ like helicase 5 RECQL5 RECQ5 7.6kb Anti-sense Chr. 17 - 75626845 - 75667202 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008912.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000577342.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000577785.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000578300.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000578960.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000610510.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000612587.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000619501.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000621743.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000633867.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 NM_001309242.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 TC1700008912.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000577342.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000577785.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000578300.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000578960.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000610510.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000612587.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000619501.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000621743.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 ENST00000633867.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166934.hg.1 NM_001309242.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0