Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004820.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004820.hg.1 |
ENST00000374480.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004820.hg.1 |
ENST00000374495.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004820.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004820.hg.1 |
TC0X000374.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
TC0X00007535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00151601.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000307959.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000374495.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000477379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000480877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
ENST00000620997.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007535.hg.1 |
NM_004312.2 |
Refseq |