Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01021953_m1
          See other TIAM1 GE Assays ›
          Gene Symbol:
          TIAM1
          Gene Name:
          T-cell lymphoma invasion and metastasis 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 31118418 - 31559977 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01021953_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TIAM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003253.2 NP_003244.2 26 - 27 4516 62
          XM_005261037.1 XP_005261094.1 25 - 26 4649 62
          XM_005261038.2 XP_005261095.1 24 - 25 4383 62
          XM_005261040.2 XP_005261097.1 25 - 26 4649 62
          XM_011529711.1 XP_011528013.1 26 - 27 4600 62
          XM_011529712.1 XP_011528014.1 26 - 27 4524 62
          XM_011529713.1 XP_011528015.1 26 - 27 4934 62
          XM_017028447.1 XP_016883936.1 26 - 27 4703 62
          XM_017028448.1 XP_016883937.1 26 - 27 4673 62
          XM_017028449.1 XP_016883938.1 25 - 26 4580 62
          XM_017028450.1 XP_016883939.1 25 - 26 4600 62
          XM_017028451.1 XP_016883940.1 25 - 26 5080 62
          XM_017028452.1 XP_016883941.1 27 - 28 4880 62
          XM_017028453.1 XP_016883942.1 25 - 26 4555 62
          GenBank mRNA AB209101.1 - 22 - 23 3530 62
          BC117192.1 - 23 - 24 4091 40125743 62
          BC117196.1 - 23 - 24 4091 40125747 62
          BC143970.1 - 23 - 24 4091 62
          BC143980.1 - 21 - 22 3911 9052498 62
          U16296.1 - 26 - 27 4517 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100008555.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000388.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TIAM1

          Gene Ontology Categories:

          Function(s) Process(es)

          cardiac muscle hypertrophy
          apoptotic process
          cell-matrix adhesion
          cell migration
          Rac protein signal transduction
          positive regulation of protein binding
          regulation of Rho protein signal transduction
          response to cocaine
          positive regulation of apoptotic process
          positive regulation of JUN kinase activity
          ephrin receptor signaling pathway
          positive regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          Wnt signaling pathway, planar cell polarity pathway
          positive regulation of dendritic spine morphogenesis
          regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of ERK1 and ERK2 cascade
          protein localization to membrane
          activation of GTPase activity
          positive regulation of Schwann cell chemotaxis
          regulation of dopaminergic neuron differentiation
          neuron projection extension
          regulation of non-canonical Wnt signaling pathway
          guanyl-nucleotide exchange factor activity
          protein binding
          microtubule binding
          lipid binding
          kinase binding
          receptor tyrosine kinase binding
          small GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21004263.hg.1 TC21000388.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21004263.hg.1 ENST00000286827.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21004263.hg.1 ENST00000399841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21004263.hg.1 ENST00000541036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21004263.hg.1 NM_003253.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000286827.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000423206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000491927.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000541036.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 NM_003253.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 TC2100008555.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000541036.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000286827.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000423206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 ENST00000491927.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100158765.hg.1 NM_003253.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0