Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032119.3 | NP_115495.3 | 50 - 51 | 10646 | 100 | |
NR_003149.1 | 50 - 51 | 10563 | 100 | |||
XM_017009963.1 | XP_016865452.1 | 50 - 51 | 10786 | 100 | ||
XM_017009964.1 | XP_016865453.1 | 50 - 51 | 10783 | 100 | ||
XM_017009965.1 | XP_016865454.1 | 50 - 51 | 10668 | 100 | ||
XM_017009966.1 | XP_016865455.1 | 49 - 50 | 10705 | 100 | ||
XM_017009967.1 | XP_016865456.1 | 49 - 50 | 10575 | 100 | ||
XM_017009968.1 | XP_016865457.1 | 50 - 51 | 10786 | 100 | ||
XM_017009969.1 | XP_016865458.1 | 50 - 51 | 10786 | 100 | ||
XM_017009970.1 | XP_016865459.1 | 50 - 51 | 10786 | 100 | ||
XM_017009971.1 | XP_016865460.1 | 50 - 51 | 10786 | 100 | ||
XM_017009972.1 | XP_016865461.1 | 22 - 23 | 4068 | 100 | ||
XM_017009973.1 | XP_016865462.1 | 27 - 28 | 5279 | 100 | ||
XM_017009974.1 | XP_016865463.1 | 50 - 51 | 10786 | 100 | ||
GenBank mRNA | AL136541.1 | - | 18 - 19 | 3271 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |