Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
ENST00000239472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
ENST00000308284.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
ENST00000414025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
ENST00000466953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
ENST00000495165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
NM_033127.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055140.hg.1 |
TC01003560.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000308284.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000327037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000495165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000528461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
NM_033127.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
TC0100018537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000308284.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000327037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000495165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
ENST00000528461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244139.hg.1 |
NM_033127.2 |
Refseq |