Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01024525_g1
          See other HYOU1 GE Assays ›
          Gene Symbol:
          HYOU1
          Gene Name:
          hypoxia up-regulated 1
          Gene Aliases:
          GRP-170, Grp170, HSP12A, ORP-150, ORP150
          Chromosome Location:
          Chr.11: 119044185 - 119057246 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01024525_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HYOU1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130991.2 NP_001124463.1 5 - 6 521 59
          NM_006389.4 NP_006380.1 5 - 6 654 59
          XM_005271392.3 XP_005271449.1 5 - 6 515 59
          XM_005271393.3 XP_005271450.1 5 - 6 495 59
          XM_005271394.3 XP_005271451.1 5 - 6 562 59
          XM_017017095.1 XP_016872584.1 5 - 6 550 59
          XM_017017096.1 XP_016872585.1 5 - 6 571 59
          XM_017017097.1 XP_016872586.1 5 - 6 494 59
          GenBank mRNA AK127324.1 - 4 - 5 523 59
          AK222756.1 - 5 - 6 547 59
          AK294905.1 - 6 - 7 682 59
          AK299648.1 - 5 - 6 477 59
          AK301514.1 - 3 - 4 213 59
          AK304264.1 - 4 - 5 453 59
          AK314178.1 - 5 - 6 545 59
          BC017726.1 - 5 - 6 472 59
          BC072436.1 - 5 - 6 520 5756154 59
          BP236985.1 - 5 - 6 489 59
          DQ350134.1 - 5 - 6 542 59
          DQ372932.1 - 5 - 6 542 59
          U65785.1 - 5 - 6 522 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC11002361.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK301514.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HYOU1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to ischemia
          endoplasmic reticulum to Golgi vesicle-mediated transport
          receptor-mediated endocytosis
          response to endoplasmic reticulum stress
          IRE1-mediated unfolded protein response
          cellular response to hypoxia
          negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
          negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway
          ATP binding
          chaperone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000353883.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000404233.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000525859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000529972.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000535579.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000536103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000543287.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 ENST00000544701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 NM_001130991.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 NM_006389.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028403.hg.1 TC11002361.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000353883.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000404233.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000525859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000526571.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000529972.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000535579.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000536103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000541069.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000543287.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 ENST00000544701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 NM_001130991.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 NM_006389.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11028405.hg.1 TC11002361.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline