Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01026941_m1
          See other KTN1 GE Assays ›
          Gene Symbol:
          KTN1
          Gene Name:
          kinectin 1
          Gene Aliases:
          CG1, KNT, MU-RMS-40.19
          Chromosome Location:
          Chr.14: 55580207 - 55684584 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01026941_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KTN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079521.1 NP_001072989.1 13 - 14 2004 95
          NM_001079522.1 NP_001072990.1 14 - 15 2128 95
          NM_001271014.1 NP_001257943.1 14 - 15 2128 95
          NM_004986.3 NP_004977.2 13 - 14 2004 95
          NR_073128.1 14 - 15 2128 95
          NR_073129.1 14 - 15 2128 95
          XM_006720138.2 XP_006720201.1 14 - 15 2119 95
          XM_006720139.2 XP_006720202.1 13 - 14 1967 95
          XM_006720140.3 XP_006720203.1 13 - 14 1967 95
          XM_006720141.2 XP_006720204.1 13 - 14 1967 95
          XM_011536751.2 XP_011535053.1 13 - 14 2225 95
          XM_011536752.1 XP_011535054.1 13 - 14 1967 95
          XM_011536753.1 XP_011535055.1 13 - 14 1967 95
          XM_017021278.1 XP_016876767.1 13 - 14 7817 95
          XM_017021279.1 XP_016876768.1 13 - 14 1967 95
          XM_017021280.1 XP_016876769.1 14 - 15 2119 95
          XM_017021281.1 XP_016876770.1 14 - 15 7941 95
          XM_017021282.1 XP_016876771.1 14 - 15 2122 95
          XM_017021283.1 XP_016876772.1 14 - 15 2056 95
          XM_017021284.1 XP_016876773.1 13 - 14 1967 95
          XM_017021285.1 XP_016876774.1 14 - 15 2099 95
          XM_017021286.1 XP_016876775.1 13 - 14 1967 95
          GenBank mRNA AK302797.1 - 14 - 15 2082 95
          AY264265.1 - 14 - 15 2013 95
          AY536375.1 - 3 - 4 217 95
          BC050555.1 - 13 - 14 1943 95
          BC112337.1 - 14 - 15 2011 40068009 95
          BC117132.1 - 13 - 14 1945 40125683 95
          BC143720.1 - 13 - 14 1945 9052234 95
          D13629.1 - 13 - 14 1909 95
          L25616.1 - 12 - 13 1866 95
          Z22551.1 - 13 - 14 1892 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400007244.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000326.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KTN1

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          cell-cell adhesion
          RNA binding
          protein binding
          kinesin binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000395308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000395309.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000395311.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000395314.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000413890.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000416613.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 ENST00000438792.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 NM_001079521.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 NM_001079522.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 NM_004986.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14003510.hg.1 TC14000326.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395308.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395309.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395311.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395314.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000413890.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000438792.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000459737.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001079521.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001079522.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001271014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_004986.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NR_073128.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NR_073129.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 TC1400007244.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395308.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395309.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395311.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000395314.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000413890.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000438792.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 ENST00000459737.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001079521.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001079522.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_001271014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NM_004986.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NR_073128.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400150208.hg.1 NR_073129.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0