Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
ENST00000238936.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
ENST00000366277.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
ENST00000369930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
ENST00000447593.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009006.hg.1 |
TC10000757.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159786.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159786.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159786.hg.1 |
TC1000008733.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000159786.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159786.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000238936.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000369931.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000428200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000450947.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
ENST00000469294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008733.hg.1 |
NM_024789.3 |
Refseq |