Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000311027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000342922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000349238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000395344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000402192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000402799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
ENST00000407859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_001135943.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_001135944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_003682.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_130470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_130471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_130472.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_130473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
NM_130476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005280.hg.1 |
TC11000406.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000311027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000342922.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000349238.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000395344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000402192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000402799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000407859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000460452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_001135943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_001135944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_003682.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130472.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
TC1100007473.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000349238.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000311027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000342922.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000395344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000402192.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000402799.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000407859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
ENST00000460452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_001135943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_001135944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_003682.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130472.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153267.hg.1 |
NM_130476.2 |
Refseq |