Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01027760_m1
          See other MADD GE Assays ›
          Gene Symbol:
          MADD
          Gene Name:
          MAP kinase activating death domain
          Gene Aliases:
          DENN, IG20, RAB3GEP
          Chromosome Location:
          Chr.11: 47269376 - 47330031 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01027760_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MADD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135943.1 NP_001129415.1 31 - 32 4701 83
          NM_001135944.1 NP_001129416.1 31 - 32 4668 83
          NM_003682.3 NP_003673.3 34 - 35 4986 83
          NM_130470.2 NP_569826.2 31 - 32 4975 83
          NM_130471.2 NP_569827.2 32 - 33 4740 83
          NM_130472.2 NP_569828.2 31 - 32 4680 83
          NM_130473.2 NP_569829.2 32 - 33 4869 83
          NM_130476.2 NP_569832.2 31 - 32 4830 83
          XM_005253189.2 XP_005253246.1 34 - 35 5010 83
          XM_005253196.2 XP_005253253.1 32 - 33 4893 83
          XM_005253199.2 XP_005253256.1 31 - 32 4833 83
          XM_005253201.2 XP_005253258.1 32 - 33 4764 83
          XM_005253203.2 XP_005253260.1 31 - 32 4704 83
          XM_005253204.2 XP_005253261.1 31 - 32 4692 83
          XM_017018478.1 XP_016873967.1 34 - 35 4998 83
          XM_017018479.1 XP_016873968.1 33 - 34 4944 83
          XM_017018480.1 XP_016873969.1 32 - 33 4890 83
          XM_017018481.1 XP_016873970.1 32 - 33 4884 83
          XM_017018482.1 XP_016873971.1 32 - 33 4881 83
          XM_017018483.1 XP_016873972.1 34 - 35 4869 83
          XM_017018484.1 XP_016873973.1 31 - 32 4821 83
          XM_017018485.1 XP_016873974.1 31 - 32 4782 83
          XM_017018486.1 XP_016873975.1 33 - 34 4815 83
          XM_017018487.1 XP_016873976.1 33 - 34 4809 83
          GenBank mRNA AF440100.1 - 34 - 35 4960 83
          AF440101.1 - 32 - 33 4843 83
          AF440102.1 - 32 - 33 4714 83
          AF440103.1 - 31 - 32 4654 83
          BC040484.1 - 31 - 32 4685 5271798 83
          U44953.1 - 31 - 32 4794 83
          U77352.1 - 31 - 32 4943 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC11000406.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MADD

          Gene Ontology Categories:

          Function(s) Process(es)

          cell surface receptor signaling pathway
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of Rab protein signal transduction
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of cell cycle
          execution phase of apoptosis
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of extrinsic apoptotic signaling pathway
          death receptor binding
          protein binding
          protein kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000311027.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000342922.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000349238.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000395336.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000395344.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000402192.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000402799.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000406482.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 ENST00000407859.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_001135943.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_001135944.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_003682.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130470.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130471.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130472.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130474.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 NM_130476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11005283.hg.1 TC11000406.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0