Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01031017_m1
See other ZMYND8 GE Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01031017_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001281773.2
NP_001268702.1
22 - 23
3771
64
NM_001281774.2
NP_001268703.1
22 - 23
3633
64
NM_001281775.2
NP_001268704.1
21 - 22
3700
64
NM_001281776.2
NP_001268705.1
20 - 21
3319
64
NM_001281777.2
NP_001268706.1
21 - 22
3487
64
NM_001281779.2
NP_001268708.1
22 - 23
3693
64
NM_001281782.2
NP_001268711.1
20 - 21
3406
64
NM_183047.3
NP_898868.1
21 - 22
3562
64
XM_005260356.4
XP_005260413.1
21 - 22
3737
64
XM_005260358.2
XP_005260415.1
20 - 21
3479
64
XM_005260360.4
XP_005260417.1
21 - 22
4202
64
XM_005260362.2
XP_005260419.1
21 - 22
3477
64
XM_005260364.2
XP_005260421.1
21 - 22
3462
64
XM_006723760.3
XP_006723823.1
21 - 22
3875
64
XM_006723762.3
XP_006723825.1
21 - 22
3576
64
XM_006723763.1
XP_006723826.1
21 - 22
3491
64
XM_006723766.1
XP_006723829.1
21 - 22
3435
64
XM_011528749.2
XP_011527051.1
21 - 22
4381
64
XM_011528751.1
XP_011527053.1
21 - 22
3485
64
XM_011528752.1
XP_011527054.1
19 - 20
3326
64
XM_011528753.1
XP_011527055.1
22 - 23
3496
64
XM_011528754.1
XP_011527056.1
21 - 22
3423
64
XM_017027761.1
XP_016883250.1
21 - 22
3948
64
XM_017027762.1
XP_016883251.1
23 - 24
4715
64
XM_017027766.1
XP_016883255.1
21 - 22
3477
64
XM_017027774.1
XP_016883263.1
20 - 21
3455
64
XM_017027775.1
XP_016883264.1
21 - 22
3285
64
XM_017027779.1
XP_016883268.1
18 - 19
2576
64
GenBank mRNA
AK026151.1
-
11 - 12
2107
64
AK122966.1
-
20 - 21
3183
64
AK225857.1
-
21 - 22
3508
64
AK295747.1
-
21 - 22
3584
64
AK299899.1
-
22 - 23
3583
64
BC030721.1
-
18 - 19
3062
64
BX641005.1
-
22 - 23
3578
64
DQ082998.1
-
20 - 21
3481
64
DQ082999.1
-
19 - 20
3100
64
DQ368669.1
-
20 - 21
3343
64
DQ368672.1
-
20 - 21
3268
64
DQ368673.1
-
20 - 21
3481
64
DQ368675.1
-
21 - 22
3474
64
DQ368678.1
-
19 - 20
3187
64
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ZMYND8
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II transcription regulatory region sequence-specific DNA binding
chromatin binding
transcription coregulator activity
transcription corepressor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
DNA-binding transcription factor binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-85f78c8899-kf4dt:80/100.66.130.62:80.
git-commit: 7f1fbef35c8161921da6e3861d81379c0ca55d8f
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix