Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01031358_m1
          See other MYCBP2 GE Assays ›
          Gene Symbol:
          MYCBP2
          Gene Name:
          MYC binding protein 2, E3 ubiquitin protein ligase
          Gene Aliases:
          Myc-bp2, PAM, Phr
          Chromosome Location:
          Chr.13: 77044657 - 77327066 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01031358_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYCBP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015057.4 NP_055872.4 36 - 37 5733 102
          XM_005266299.2 XP_005266356.1 36 - 37 5757 102
          XM_006719779.2 XP_006719842.1 36 - 37 5757 102
          XM_006719780.2 XP_006719843.1 36 - 37 5757 102
          XM_006719781.2 XP_006719844.1 36 - 37 5757 102
          XM_006719782.2 XP_006719845.1 36 - 37 5757 102
          XM_006719783.2 XP_006719846.1 36 - 37 5757 102
          XM_006719784.2 XP_006719847.1 35 - 36 5649 102
          XM_006719785.2 XP_006719848.1 36 - 37 5757 102
          XM_006719786.2 XP_006719849.1 36 - 37 5757 102
          XM_006719787.2 XP_006719850.1 36 - 37 5757 102
          XM_006719788.2 XP_006719851.1 36 - 37 5757 102
          XM_006719789.2 XP_006719852.1 36 - 37 5757 102
          XM_011535004.2 XP_011533306.1 35 - 36 5296 102
          XM_011535005.2 XP_011533307.1 19 - 20 2837 102
          XM_017020454.1 XP_016875943.1 36 - 37 5757 102
          XM_017020455.1 XP_016875944.1 36 - 37 5757 102
          XM_017020456.1 XP_016875945.1 36 - 37 5757 102
          XM_017020457.1 XP_016875946.1 36 - 37 5757 102
          XM_017020458.1 XP_016875947.1 35 - 36 5649 102
          XM_017020459.1 XP_016875948.1 35 - 36 5649 102
          XM_017020460.1 XP_016875949.1 36 - 37 5757 102
          XM_017020461.1 XP_016875950.1 36 - 37 5757 102
          XM_017020462.1 XP_016875951.1 36 - 37 5757 102
          XM_017020463.1 XP_016875952.1 35 - 36 5649 102
          XM_017020464.1 XP_016875953.1 36 - 37 5757 102
          XM_017020465.1 XP_016875954.1 35 - 36 5649 102
          XM_017020466.1 XP_016875955.1 36 - 37 5757 102
          XM_017020467.1 XP_016875956.1 36 - 37 5757 102
          XM_017020468.1 XP_016875957.1 35 - 36 5244 102
          GenBank mRNA AF075587.1 - 37 - 38 5498 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009293.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000744.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYCBP2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein ubiquitination
          protein binding
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 ENST00000357337.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 ENST00000360084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 ENST00000407578.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 ENST00000544440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 NM_015057.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008611.hg.1 TC13000744.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 ENST00000357337.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 ENST00000544440.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 NM_015057.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 TC1300009293.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 NM_015057.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 ENST00000357337.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162418.hg.1 ENST00000544440.6 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0