Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01031614_m1
          See other SLC9A3R1 GE Assays ›
          Gene Symbol:
          SLC9A3R1
          Gene Name:
          SLC9A3 regulator 1
          Gene Aliases:
          EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2
          Chromosome Location:
          Chr.17: 74748612 - 74769360 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01031614_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC9A3R1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004252.4 NP_004243.1 2 - 3 841 53
          GenBank mRNA AF015926.1 - 2 - 3 818 53
          AF036241.1 - 2 - 3 776 53
          AK026581.1 - 2 - 3 824 53
          AK094467.1 - 3 - 4 675 53
          AK128474.1 - 3 - 4 790 53
          AK312498.1 - 2 - 3 857 53
          BC001443.2 - 2 - 3 794 3138881 53
          BC003361.1 - 2 - 3 811 2900516 53
          BC011777.1 - 2 - 3 810 3640162 53
          BC049220.1 - 2 - 3 817 53
          BC053350.1 - 2 - 3 801 5218414 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008867.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000834.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC9A3R1

          Gene Ontology Categories:

          Function(s) Process(es)

          renal sodium ion transport
          nuclear migration
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of sound
          negative regulation of cell population proliferation
          regulation of cell shape
          regulation of cell size
          negative regulation of platelet-derived growth factor receptor signaling pathway
          negative regulation of phosphatidylinositol 3-kinase signaling
          Wnt signaling pathway
          cAMP-mediated signaling
          gland morphogenesis
          microvillus assembly
          actin cytoskeleton organization
          negative regulation of cell migration
          cellular phosphate ion homeostasis
          regulation of sodium:proton antiporter activity
          negative regulation of sodium:proton antiporter activity
          bile acid secretion
          glutathione transport
          regulation of excretion
          establishment of epithelial cell apical/basal polarity
          regulation of protein kinase activity
          negative regulation of mitotic cell cycle
          establishment of Golgi localization
          negative regulation of protein kinase B signaling
          auditory receptor cell stereocilium organization
          phospholipase C-activating dopamine receptor signaling pathway
          protein-containing complex assembly
          renal absorption
          negative regulation of ERK1 and ERK2 cascade
          protein localization to plasma membrane
          negative regulation of canonical Wnt signaling pathway
          renal phosphate ion absorption
          positive regulation of intrinsic apoptotic signaling pathway
          signaling receptor binding
          protein binding
          beta-catenin binding
          chloride channel regulator activity
          phosphatase binding
          PDZ domain binding
          beta-2 adrenergic receptor binding
          protein self-association
          protein-containing complex binding
          dopamine receptor binding
          molecular adaptor activity
          growth factor receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000581999.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700008867.hg.1 NR_037409.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010803.hg.1 ENST00000262613.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010803.hg.1 ENST00000413388.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010803.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010803.hg.1 TC17000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17010804.hg.1 ENST00000262613.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17010804.hg.1 NM_004252.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17010804.hg.1 TC17000834.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000581999.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 ENST00000583369.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay TC1700008867.hg.1 NR_037409.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 TC1700008867.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000581356.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166245.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 NM_004252.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 TC1700008867.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000262613.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000413388.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 ENST00000578958.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700166244.hg.1 NM_004252.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0