Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01037006_gH
          See other MMP14 GE Assays ›
          Gene Symbol:
          MMP14
          Gene Name:
          matrix metallopeptidase 14
          Gene Aliases:
          MMP-14, MMP-X1, MT-MMP, MT-MMP 1, MT1-MMP, MT1MMP, MTMMP1, WNCHRS
          Chromosome Location:
          Chr.14: 22836533 - 22847600 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01037006_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MMP14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004995.3 NP_004986.1 6 - 7 1295 70
          GenBank mRNA AK291325.1 - 6 - 7 1237 70
          AK312657.1 - 6 - 7 1243 70
          BC064803.1 - 6 - 7 1229 6046773 70
          HE978355.1 - 6 - 7 1244 70
          U41078.1 - 6 - 7 1010 70
          X83535.1 - 6 - 7 1129 70
          X90925.1 - 6 - 7 1122 70
          Z48481.1 - 6 - 7 1123 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400006659.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000133.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MMP14

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovarian follicle development
          response to hypoxia
          endothelial cell proliferation
          endochondral ossification
          proteolysis
          response to oxidative stress
          male gonad development
          response to mechanical stimulus
          response to hormone
          positive regulation of myotube differentiation
          positive regulation of peptidase activity
          response to organic cyclic compound
          extracellular matrix disassembly
          positive regulation of cell growth
          lung development
          positive regulation of cell migration
          collagen catabolic process
          zymogen activation
          endodermal cell differentiation
          chondrocyte proliferation
          astrocyte cell migration
          response to estrogen
          positive regulation of B cell differentiation
          negative regulation of Notch signaling pathway
          embryonic cranial skeleton morphogenesis
          branching morphogenesis of an epithelial tube
          tissue remodeling
          negative regulation of focal adhesion assembly
          craniofacial suture morphogenesis
          metalloendopeptidase activity
          serine-type endopeptidase activity
          integrin binding
          calcium ion binding
          protein binding
          zinc ion binding
          peptidase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14001149.hg.1 ENST00000311852.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14001149.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14001149.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14001149.hg.1 TC14000133.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14001150.hg.1 ENST00000311852.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14001150.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14001150.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14001150.hg.1 TC14000133.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 TC1400006659.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145531.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 TC1400006659.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400145532.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000311852.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547279.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000547596.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 ENST00000548761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq
          Affymetrix Human Clariom D Assay TC1400006659.hg.1 NM_004995.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0