Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01037202_g1
          See other KIF5B GE Assays ›
          Gene Symbol:
          KIF5B
          Gene Name:
          kinesin family member 5B
          Gene Aliases:
          HEL-S-61, KINH, KNS, KNS1, UKHC
          Chromosome Location:
          Chr.10: 32009010 - 32056443 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01037202_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIF5B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004521.2 NP_004512.1 4 - 5 860 154
          XM_017016224.1 XP_016871713.1 4 - 5 466 154
          GenBank mRNA AK289413.1 - 4 - 5 806 154
          AK307502.1 - 5 - 6 557 154
          BC010885.1 - 4 - 5 796 154
          BC065267.1 - 4 - 5 817 154
          BC080604.1 - 4 - 5 791 154
          BC111042.1 - 4 - 5 800 154
          BC126279.1 - 4 - 5 613 8991995 154
          BC126281.1 - 4 - 5 613 8991997 154
          FJ224324.1 - 4 - 5 703 154
          X65873.1 - 4 - 5 703 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010249.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001160.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KIF5B

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          cytoplasm organization
          axon guidance
          cytoskeleton-dependent intracellular transport
          positive regulation of synaptic transmission, GABAergic
          stress granule disassembly
          regulation of membrane potential
          positive regulation of potassium ion transport
          vesicle transport along microtubule
          plus-end-directed vesicle transport along microtubule
          cell-cell adhesion
          positive regulation of protein localization to plasma membrane
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          plus-end-directed microtubule motor activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10014583.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10014583.hg.1 NM_004521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10014583.hg.1 TC10001160.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000169797.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169797.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000169797.hg.1 TC1000010249.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000169797.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000169797.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000302418.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 ENST00000493889.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq
          Affymetrix Human Clariom D Assay TC1000010249.hg.1 NM_004521.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0