Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016514.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016514.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10016514.hg.1 |
TC10001317.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000491922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010725.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
NM_005436.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
TC1000010725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
ENST00000263102.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
ENST00000518638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000173107.hg.1 |
NM_005436.4 |
Refseq |