Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01038438_m1
          See other RARS2 GE Assays ›
          Gene Symbol:
          RARS2
          Gene Name:
          arginyl-tRNA synthetase 2, mitochondrial
          Gene Aliases:
          ArgRS, DALRD2, PCH6, PRO1992, RARSL
          Chromosome Location:
          Chr.6: 87513744 - 87590032 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01038438_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RARS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318785.1 NP_001305714.1 9 - 10 1122 85
          NM_020320.4 NP_064716.2 10 - 11 957 85
          NR_134857.1 10 - 11 957 85
          XM_011535948.2 XP_011534250.1 10 - 11 942 85
          XM_011535949.2 XP_011534251.1 10 - 11 942 85
          XM_017011072.1 XP_016866561.1 10 - 11 1123 85
          XM_017011073.1 XP_016866562.1 11 - 12 1341 85
          XM_017011074.1 XP_016866563.1 10 - 11 944 85
          XM_017011075.1 XP_016866564.1 9 - 10 1067 85
          XM_017011076.1 XP_016866565.1 11 - 12 1285 85
          XM_017011077.1 XP_016866566.1 9 - 10 844 85
          XM_017011078.1 XP_016866567.1 9 - 10 888 85
          XM_017011079.1 XP_016866568.1 10 - 11 1123 85
          XM_017011080.1 XP_016866569.1 11 - 12 1285 85
          XM_017011081.1 XP_016866570.1 9 - 10 888 85
          GenBank mRNA AK023550.1 - 10 - 11 900 85
          AK093934.1 - 10 - 11 1330 85
          AK315669.1 - 10 - 11 894 85
          BC010420.1 - 10 - 11 3613406 85
          BC105784.1 - 9 - 10 1061 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012512.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001928.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RARS2

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA aminoacylation for protein translation
          arginyl-tRNA aminoacylation
          mitochondrial translation
          RNA binding
          arginine-tRNA ligase activity
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06023626.hg.1 ENST00000369523.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023626.hg.1 ENST00000369536.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023626.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06023626.hg.1 TC06001928.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186192.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600186192.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186192.hg.1 TC0600012512.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600186192.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600186192.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000369536.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000451155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000493269.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 ENST00000497828.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq
          Affymetrix Human Clariom D Assay TC0600012512.hg.1 NM_020320.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0