Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01038919_m1
          See other PIKFYVE GE Assays ›
          Gene Symbol:
          PIKFYVE
          Gene Name:
          phosphoinositide kinase, FYVE-type zinc finger containing
          Gene Aliases:
          CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
          Chromosome Location:
          Chr.2: 208266178 - 208358751 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01038919_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIKFYVE
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015040.3 NP_055855.2 39 - 40 6002 154
          XM_011510778.2 XP_011509080.1 40 - 41 6052 154
          XM_011510779.2 XP_011509081.1 40 - 41 5985 154
          XM_011510780.2 XP_011509082.1 40 - 41 6070 154
          XM_011510781.2 XP_011509083.1 40 - 41 5993 154
          XM_011510782.2 XP_011509084.1 39 - 40 5887 154
          XM_011510783.2 XP_011509085.1 39 - 40 5884 154
          XM_011510784.2 XP_011509086.1 39 - 40 5902 154
          XM_011510785.2 XP_011509087.1 39 - 40 5824 154
          XM_011510786.2 XP_011509088.1 38 - 39 5719 154
          XM_011510787.1 XP_011509089.1 38 - 39 5728 154
          XM_011510788.1 XP_011509090.1 37 - 38 5711 154
          XM_011510789.2 XP_011509091.1 36 - 37 5533 154
          XM_011510790.1 XP_011509092.1 38 - 39 5830 154
          XM_017003568.1 XP_016859057.1 39 - 40 5888 154
          XM_017003569.1 XP_016859058.1 38 - 39 5720 154
          XM_017003570.1 XP_016859059.1 36 - 37 5569 154
          XM_017003571.1 XP_016859060.1 35 - 36 5365 154
          XM_017003572.1 XP_016859061.1 38 - 39 5706 154
          XM_017003573.1 XP_016859062.1 35 - 36 5414 154
          XM_017003574.1 XP_016859063.1 34 - 35 5085 154
          GenBank mRNA AB023198.1 - 12 - 13 1285 154
          AK091482.1 - 20 - 21 2738 154
          AY457063.1 - 39 - 40 6006 154
          EU794609.1 - 39 - 40 6006 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010624.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001235.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIKFYVE

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          receptor-mediated endocytosis
          myelin assembly
          protein localization to nucleus
          intracellular signal transduction
          phosphatidylinositol-3-phosphate biosynthetic process
          retrograde transport, endosome to Golgi
          phosphatidylinositol phosphate biosynthetic process
          phosphatidylinositol 5-phosphate metabolic process
          regulation of autophagosome assembly
          1-phosphatidylinositol-3-phosphate 5-kinase activity
          protein binding
          ATP binding
          zinc ion binding
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02018945.hg.1 ENST00000264380.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018945.hg.1 ENST00000392200.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018945.hg.1 NM_015040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018945.hg.1 TC02001235.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200175305.hg.1 ENST00000264380.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200175305.hg.1 NM_015040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200175305.hg.1 TC0200010624.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200175305.hg.1 ENST00000264380.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200175305.hg.1 NM_015040.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0