Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01039877_m1
          See other MAP3K5 GE Assays ›
          Gene Symbol:
          MAP3K5
          Gene Name:
          mitogen-activated protein kinase kinase kinase 5
          Gene Aliases:
          ASK1, MAPKKK5, MEKK5
          Chromosome Location:
          Chr.6: 136557046 - 136793098 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01039877_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP3K5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005923.3 NP_005914.1 11 - 12 2148 79
          XM_011535839.2 XP_011534141.2 12 - 13 2472 79
          XM_017010870.1 XP_016866359.1 12 - 13 2471 79
          XM_017010871.1 XP_016866360.1 12 - 13 2471 79
          XM_017010872.1 XP_016866361.1 12 - 13 2470 79
          XM_017010873.1 XP_016866362.1 12 - 13 2469 79
          XM_017010876.1 XP_016866365.1 10 - 11 1479 79
          XM_017010877.1 XP_016866366.1 10 - 11 1716 79
          GenBank mRNA AB209093.1 - 11 - 12 2252 79
          AK293953.1 - 11 - 12 1408 79
          AK294507.1 - 2 - 3 264 79
          BC054503.2 - 11 - 12 2095 6143181 79
          BC088829.1 - 11 - 12 2149 6007002 79
          D84476.2 - 11 - 12 2054 79
          U67156.1 - 11 - 12 2147 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013280.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002146.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK294507.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAP3K5

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          response to ischemia
          protein phosphorylation
          JNK cascade
          intrinsic apoptotic signaling pathway in response to oxidative stress
          viral process
          response to endoplasmic reticulum stress
          p38MAPK cascade
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of JUN kinase activity
          innate immune response
          positive regulation of myoblast differentiation
          positive regulation of JNK cascade
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to hydrogen peroxide
          apoptotic signaling pathway
          programmed necrotic cell death
          positive regulation of neuron death
          cellular response to reactive nitrogen species
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          protein binding
          ATP binding
          protein kinase binding
          protein phosphatase binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027072.hg.1 ENST00000355845.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027072.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027072.hg.1 ENST00000367768.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06027072.hg.1 NM_005923.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06027072.hg.1 TC06002146.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600191950.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600191950.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600191950.hg.1 TC0600013280.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600191950.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600191950.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000359015.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 ENST00000463140.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq
          Affymetrix Human Clariom D Assay TC0600013280.hg.1 NM_005923.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0