Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045928.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045928.hg.1 |
ENST00000543700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045928.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045928.hg.1 |
TC02002847.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045927.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045927.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045927.hg.1 |
TC02002847.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217362.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217362.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217362.hg.1 |
TC0200015958.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200217362.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217362.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000341772.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
ENST00000482831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015958.hg.1 |
NM_139072.3 |
Refseq |