Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01040134_m1
          See other PINK1 GE Assays ›
          Gene Symbol:
          PINK1
          Gene Name:
          PTEN induced putative kinase 1
          Gene Aliases:
          BRPK, PARK6
          Chromosome Location:
          Chr.1: 20633455 - 20651511 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01040134_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PINK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032409.2 NP_115785.1 4 - 5 1045 79
          GenBank mRNA AB053323.1 - 4 - 5 1045 79
          AF316873.1 - 4 - 5 1021 79
          AK130746.1 - 2 - 3 1148 79
          BC009534.1 - 1 - 2 118 79
          BC028215.1 - 4 - 5 960 5214483 79

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PINK1

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy of mitochondrion
          positive regulation of protein phosphorylation
          regulation of oxidative phosphorylation
          protein phosphorylation
          ubiquitin-dependent protein catabolic process
          response to stress
          response to oxidative stress
          mitochondrion organization
          regulation of hydrogen peroxide metabolic process
          negative regulation of gene expression
          regulation of mitochondrion organization
          positive regulation of peptidase activity
          macroautophagy
          positive regulation of macroautophagy
          negative regulation of macroautophagy
          phosphorylation
          protein ubiquitination
          peptidyl-serine phosphorylation
          respiratory electron transport chain
          regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          activation of protein kinase B activity
          positive regulation of synaptic transmission, dopaminergic
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of dopamine secretion
          cellular response to oxidative stress
          positive regulation of protein dephosphorylation
          intracellular signal transduction
          peptidyl-serine autophosphorylation
          TORC2 signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein-containing complex assembly
          negative regulation of neuron apoptotic process
          positive regulation of translation
          negative regulation of JNK cascade
          protein stabilization
          positive regulation of DNA-binding transcription factor activity
          positive regulation of ubiquitin-protein transferase activity
          regulation of mitochondrial membrane potential
          positive regulation of protein kinase B signaling
          regulation of proteasomal protein catabolic process
          cellular response to hypoxia
          establishment of protein localization to mitochondrion
          maintenance of protein location in mitochondrion
          positive regulation of mitochondrial fission
          positive regulation of release of cytochrome c from mitochondria
          negative regulation of mitochondrial fission
          cellular response to toxic substance
          positive regulation of mitophagy in response to mitochondrial depolarization
          regulation of synaptic vesicle transport
          negative regulation of autophagosome assembly
          positive regulation of mitochondrial electron transport, NADH to ubiquinone
          regulation of autophagy of mitochondrion
          negative regulation of autophagy of mitochondrion
          negative regulation of oxidative stress-induced cell death
          negative regulation of oxidative stress-induced neuron death
          regulation of protein targeting to mitochondrion
          negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
          negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide
          positive regulation of cristae formation
          positive regulation of protein targeting to mitochondrion
          positive regulation of free ubiquitin chain polymerization
          regulation of reactive oxygen species metabolic process
          negative regulation of reactive oxygen species metabolic process
          positive regulation of ATP biosynthetic process
          magnesium ion binding
          protease binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          calcium-dependent protein kinase activity
          kinase activity
          peptidase activator activity
          ubiquitin protein ligase binding
          protein kinase B binding
          C3HC4-type RING finger domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 ENST00000622012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007207.hg.1 NR_106732.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000321556.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000400490.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000492302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 ENST00000622012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NM_032409.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007207.hg.1 NR_106732.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0