Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168370.1 | NP_001161842.1 | 24 - 25 | 4906 | 66 | |
NM_014780.4 | NP_055595.2 | 24 - 25 | 4899 | 66 | ||
XM_005249503.2 | XP_005249560.1 | 24 - 25 | 4762 | 66 | ||
XM_017011534.1 | XP_016867023.1 | 24 - 25 | 4884 | 66 | ||
XM_017011537.1 | XP_016867026.1 | 24 - 25 | 5000 | 66 | ||
GenBank mRNA | AK292806.1 | - | 24 - 25 | 4836 | 66 | |
AK302668.1 | - | 24 - 25 | 4843 | 66 | ||
AK307936.1 | - | 14 - 15 | 2327 | 66 | ||
BC033647.1 | - | 24 - 25 | 4881 | 5580027 | 66 | |
D38548.1 | - | 24 - 25 | 4653 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |