Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01041498_m1
          See other MYBPC1 GE Assays ›
          Gene Symbol:
          MYBPC1
          Gene Name:
          myosin binding protein C, slow type
          Gene Aliases:
          LCCS4, MYBPCC, MYBPCS
          Chromosome Location:
          Chr.12: 101594849 - 101695841 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01041498_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYBPC1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001254719.1 NP_001241648.1 20 - 21 2423 87
          NM_001254720.1 NP_001241649.1 19 - 20 2387 87
          NM_001254721.1 NP_001241650.1 19 - 20 2366 87
          NM_001254722.1 NP_001241651.1 18 - 19 2345 87
          NM_001254723.1 NP_001241652.1 19 - 20 2384 87
          NM_002465.3 NP_002456.2 22 - 23 2498 87
          NM_206819.2 NP_996555.1 22 - 23 2498 87
          NM_206821.2 NP_996557.1 20 - 21 2423 87
          XM_005268876.4 XP_005268933.1 21 - 22 2432 87
          XM_006719408.3 XP_006719471.1 22 - 23 2359 87
          XM_017019317.1 XP_016874806.1 20 - 21 2385 87
          XM_017019318.1 XP_016874807.1 21 - 22 2302 87
          XM_017019321.1 XP_016874810.1 19 - 20 2328 87
          XM_017019322.1 XP_016874811.1 17 - 18 2370 87
          GenBank mRNA AK292976.1 - 22 - 23 2458 87
          AK295727.1 - 18 - 19 2305 87
          AK303401.1 - 19 - 20 2344 87
          AL832000.2 - 22 - 23 2433 87
          AL832332.2 - 22 - 23 2433 87
          AL832350.2 - 22 - 23 2433 87
          AL832354.2 - 22 - 23 2436 87
          AL832456.2 - 22 - 23 2433 87
          BC092418.1 - 20 - 21 2370 30529254 87
          BC117217.1 - 18 - 19 2280 40125768 87
          BC143495.1 - 19 - 20 2301 9052003 87
          BC143502.1 - 18 - 19 2280 9052010 87
          BC143503.1 - 20 - 21 2358 9052011 87
          BC143504.1 - 19 - 20 2322 9052012 87
          X73114.1 - 20 - 21 2364 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC12000787.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYBPC1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          muscle filament sliding
          positive regulation of protein targeting to mitochondrion
          actin binding
          protein binding
          structural constituent of muscle
          titin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000360610.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000361466.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000361685.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000392934.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000441232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000452455.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000536007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000540770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000541119.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000545503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000547405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000547509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000549145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000550270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000551300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 ENST00000553190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254720.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254721.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254722.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_001254723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_002465.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_206819.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_206820.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 NM_206821.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009987.hg.1 TC12000787.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000360610.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000361466.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000361685.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000392934.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000441232.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000452455.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000536007.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000540770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000541119.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000545503.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000547405.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000547509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000549145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000550270.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000551300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 ENST00000553190.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254718.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254720.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254721.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254722.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_001254723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_002465.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_206819.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_206820.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 NM_206821.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009986.hg.1 TC12000787.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0