Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000304076.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000539284.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000596764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000599806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000602142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_001258206.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_001258207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_005428.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
TC1900006821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000304076.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000539284.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000596764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000599806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
ENST00000602142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_001258206.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_001258207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900147936.hg.1 |
NM_005428.3 |
Refseq |