Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01041931_g1
          See other ADAM17 GE Assays ›
          Gene Symbol:
          ADAM17
          Gene Name:
          ADAM metallopeptidase domain 17
          Gene Aliases:
          ADAM18, CD156B, CSVP, NISBD, NISBD1, TACE
          Chromosome Location:
          Chr.2: 9488486 - 9555820 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01041931_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADAM17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003183.5 NP_003174.3 9 - 10 1311 94
          XM_011510375.2 XP_011508677.1 9 - 10 1330 94
          XM_011510376.2 XP_011508678.1 9 - 10 1299 94
          XM_011510377.2 XP_011508679.1 4 - 5 684 94
          XM_017004785.1 XP_016860274.1 9 - 10 1332 94
          XM_017004786.1 XP_016860275.1 8 - 9 1024 94
          GenBank mRNA BC136783.1 - 9 - 10 1173 9020773 94
          GQ402545.1 - 9 - 10 1279 94
          U69611.1 - 9 - 10 1210 94
          U69612.1 - 9 - 10 1218 94
          U86755.1 - 9 - 10 1259 94
          U92649.1 - 9 - 10 1260 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200011688.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001558.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ADAM17

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          neutrophil mediated immunity
          germinal center formation
          positive regulation of leukocyte chemotaxis
          proteolysis
          membrane protein ectodomain proteolysis
          cell adhesion
          epidermal growth factor receptor signaling pathway
          Notch signaling pathway
          Notch receptor processing
          positive regulation of cell population proliferation
          response to xenobiotic stimulus
          positive regulation of T cell chemotaxis
          B cell differentiation
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          membrane protein intracellular domain proteolysis
          response to lipopolysaccharide
          positive regulation of chemokine production
          regulation of mast cell apoptotic process
          T cell differentiation in thymus
          tumor necrosis factor-mediated signaling pathway
          cell adhesion mediated by integrin
          wound healing, spreading of epidermal cells
          positive regulation of epidermal growth factor-activated receptor activity
          spleen development
          cell motility
          defense response to Gram-positive bacterium
          positive regulation of cellular component movement
          cellular response to high density lipoprotein particle stimulus
          metalloendopeptidase activity
          Notch binding
          interleukin-6 receptor binding
          integrin binding
          protein binding
          metallopeptidase activity
          zinc ion binding
          SH3 domain binding
          PDZ domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02025243.hg.1 ENST00000310823.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025243.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02025243.hg.1 TC02001558.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000478059.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000497134.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 ENST00000618923.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay TC0200011688.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay PSR0200184954.hg.1 ENST00000310823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184954.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay PSR0200184954.hg.1 TC0200011688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184954.hg.1 NM_003183.5 Refseq
          Affymetrix Human Clariom D Assay PSR0200184954.hg.1 ENST00000310823.7 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline