Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01042400_g1
          See other MTOR GE Assays ›
          Gene Symbol:
          MTOR
          Gene Name:
          mechanistic target of rapamycin
          Gene Aliases:
          FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
          Chromosome Location:
          Chr.1: 11106531 - 11262551 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01042400_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MTOR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004958.3 NP_004949.1 35 - 36 5122 62
          XM_005263438.2 XP_005263495.1 35 - 36 5129 62
          XM_017000900.1 XP_016856389.1 32 - 33 4585 62
          XM_017000901.1 XP_016856390.1 33 - 34 4873 62
          GenBank mRNA AB209995.1 - 35 - 36 5103 62
          BC117166.1 - 34 - 35 5019 40125717 62
          L34075.1 - 35 - 36 5080 62
          U88966.1 - 35 - 36 5077 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012872.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002197.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MTOR

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          heart morphogenesis
          heart valve morphogenesis
          regulation of glycogen biosynthetic process
          energy reserve metabolic process
          'de novo' pyrimidine nucleobase biosynthetic process
          DNA repair
          protein phosphorylation
          response to stress
          signal transduction
          germ cell development
          brain development
          cell aging
          response to nutrient
          long-term memory
          visual learning
          post-embryonic development
          negative regulation of autophagy
          positive regulation of lamellipodium assembly
          positive regulation of gene expression
          positive regulation of myotube differentiation
          positive regulation of neuron maturation
          negative regulation of muscle atrophy
          cell growth
          macroautophagy
          negative regulation of macroautophagy
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          spinal cord development
          protein catabolic process
          positive regulation of actin filament polymerization
          T cell costimulation
          negative regulation of protein ubiquitination
          ruffle organization
          regulation of myelination
          cellular response to nutrient levels
          TOR signaling
          regulation of fatty acid beta-oxidation
          regulation of response to food
          response to insulin
          regulation of actin cytoskeleton organization
          social behavior
          multicellular organism growth
          growth
          wound healing
          response to cocaine
          regulation of GTPase activity
          response to amino acid
          response to morphine
          regulation of carbohydrate utilization
          positive regulation of nitric oxide biosynthetic process
          regulation of osteoclast differentiation
          positive regulation of translation
          negative regulation of cell size
          regulation of protein kinase activity
          positive regulation of transcription by RNA polymerase III
          protein autophosphorylation
          positive regulation of lipid biosynthetic process
          phosphatidylinositol-mediated signaling
          mRNA stabilization
          positive regulation of smooth muscle cell proliferation
          positive regulation of oligodendrocyte differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          voluntary musculoskeletal movement
          positive regulation of stress fiber assembly
          positive regulation of protein kinase B signaling
          cardiac muscle cell development
          cardiac muscle contraction
          maternal process involved in female pregnancy
          positive regulation of glial cell proliferation
          positive regulation of dendritic spine development
          positive regulation of cell growth involved in cardiac muscle cell development
          negative regulation of calcineurin-NFAT signaling cascade
          cellular response to hypoxia
          regulation of brown fat cell differentiation
          regulation of membrane permeability
          regulation of cellular response to heat
          positive regulation of neuron death
          positive regulation of transcription of nucleolar large rRNA by RNA polymerase I
          positive regulation of eating behavior
          positive regulation of cholangiocyte proliferation
          positive regulation of sensory perception of pain
          negative regulation of cholangiocyte apoptotic process
          positive regulation of granulosa cell proliferation
          positive regulation of skeletal muscle hypertrophy
          negative regulation of iodide transmembrane transport
          RNA polymerase III type 1 promoter sequence-specific DNA binding
          RNA polymerase III type 2 promoter sequence-specific DNA binding
          RNA polymerase III type 3 promoter sequence-specific DNA binding
          TFIIIC-class transcription factor complex binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          protein kinase binding
          protein domain specific binding
          ribosome binding
          phosphoprotein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01033950.hg.1 ENST00000361445.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033950.hg.1 ENST00000495435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033950.hg.1 NM_004958.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033950.hg.1 TC01002197.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000361445.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000476768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000495435.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 NM_004958.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 TC0100012872.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 NM_004958.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000361445.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000476768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100195134.hg.1 ENST00000495435.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0