Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01045115_m1
          See other NDRG2 GE Assays ›
          Gene Symbol:
          NDRG2
          Gene Name:
          NDRG family member 2
          Gene Aliases:
          SYLD
          Chromosome Location:
          Chr.14: 21016763 - 21070872 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01045115_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDRG2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282211.1 NP_001269140.1 5 - 6 411 77
          NM_001282212.1 NP_001269141.1 6 - 7 685 77
          NM_001282213.1 NP_001269142.1 6 - 7 690 77
          NM_001282214.1 NP_001269143.1 6 - 7 678 77
          NM_001282215.1 NP_001269144.1 6 - 7 508 77
          NM_001282216.1 NP_001269145.1 5 - 6 426 77
          NM_001320329.1 NP_001307258.1 6 - 7 1343 77
          NM_016250.2 NP_057334.1 6 - 7 533 77
          NM_201535.1 NP_963293.1 7 - 8 575 77
          NM_201536.1 NP_963294.1 5 - 6 466 77
          NM_201537.1 NP_963831.1 6 - 7 508 77
          NM_201538.1 NP_963832.1 5 - 6 443 77
          NM_201539.1 NP_963833.1 6 - 7 485 77
          NM_201540.1 NP_963834.1 7 - 8 552 77
          NM_201541.1 NP_963835.1 6 - 7 510 77
          XM_011536996.2 XP_011535298.1 7 - 8 562 77
          XM_011536997.1 XP_011535299.1 7 - 8 563 77
          XM_011536998.1 XP_011535300.1 7 - 8 553 77
          XM_011536999.1 XP_011535301.1 5 - 6 958 77
          XM_011537001.1 XP_011535303.1 6 - 7 516 77
          XM_011537002.1 XP_011535304.1 6 - 7 662 77
          XM_017021480.1 XP_016876969.1 7 - 8 514 77
          XM_017021481.1 XP_016876970.1 7 - 8 553 77
          XM_017021482.1 XP_016876971.1 7 - 8 562 77
          XM_017021483.1 XP_016876972.1 6 - 7 446 77
          XM_017021484.1 XP_016876973.1 6 - 7 488 77
          XM_017021485.1 XP_016876974.1 7 - 8 553 77
          XM_017021486.1 XP_016876975.1 7 - 8 563 77
          XM_017021487.1 XP_016876976.1 6 - 7 472 77
          XM_017021488.1 XP_016876977.1 6 - 7 513 77
          XM_017021489.1 XP_016876978.1 6 - 7 618 77
          XM_017021490.1 XP_016876979.1 5 - 6 410 77
          XM_017021491.1 XP_016876980.1 5 - 6 445 77
          XM_017021492.1 XP_016876981.1 6 - 7 516 77
          XM_017021493.1 XP_016876982.1 6 - 7 662 77
          XM_017021494.1 XP_016876983.1 7 - 8 514 77
          XM_017021495.1 XP_016876984.1 6 - 7 446 77
          XM_017021496.1 XP_016876985.1 5 - 6 445 77
          GenBank mRNA AB033074.1 - 5 - 6 395 77
          AF087872.1 - 5 - 6 449 77
          AF159092.3 - 6 - 7 456 77
          AK024521.1 - 7 - 8 508 77
          AK057843.1 - 6 - 7 479 77
          AK057847.1 - 5 - 6 443 77
          AK090657.1 - 5 - 6 382 77
          AK096999.1 - 6 - 7 1343 77
          AK130029.1 - 5 - 6 381 77
          AK293514.1 - 5 - 6 411 77
          AL136574.1 - 5 - 6 420 77
          BC010458.1 - 5 - 6 414 4215141 77
          BC011240.1 - 6 - 7 468 4153602 77
          BI759006.1 - 7 - 8 494 77
          BM810623.1 - 6 - 7 449 77
          BX247987.1 - 6 - 7 544 77
          BX248031.1 - 4 - 5 372 77
          BX647091.1 - 3 - 4 608 77
          BX647748.1 - 6 - 7 625 77
          CB153966.1 - 5 - 6 509 77
          CR749252.1 - 6 - 7 678 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC14000908.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDRG2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          signal transduction
          regulation of vascular endothelial growth factor production
          Wnt signaling pathway
          substantia nigra development
          cell differentiation
          negative regulation of smooth muscle cell proliferation
          negative regulation of ERK1 and ERK2 cascade
          regulation of platelet-derived growth factor production
          molecular_function
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000298684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000298687.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000350792.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000360463.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397844.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397847.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397851.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397853.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397855.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397856.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000397858.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000403829.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000553503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000554104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000554143.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000554277.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000554770.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000555158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 ENST00000556147.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_016250.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201537.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201538.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201539.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201540.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 NM_201541.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009819.hg.1 TC14000908.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline