Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01045159_m1
          See other CD80 GE Assays ›
          Gene Symbol:
          CD80
          Gene Name:
          CD80 molecule
          Gene Aliases:
          B7, B7-1, B7.1, BB1, CD28LG, CD28LG1, LAB7
          Chromosome Location:
          Chr.3: 119523909 - 119559709 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01045159_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005191.3 NP_005182.1 1 - 2 196 94
          XM_011513327.2 XP_011511629.1 1 - 2 174 94
          XM_017007520.1 XP_016863009.1 1 - 2 271 94
          GenBank mRNA AK292129.1 - 1 - 2 180 94
          BC042665.1 - 1 - 2 164 5181343 94
          BP225173.1 - 1 - 2 223 94
          M27533.1 - 1 - 2 118 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300012123.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001684.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK292129.1, BC042665.1, M27533.1, NM_005191.3, XM_011513327.2, XM_017007520.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD80

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell signaling
          positive regulation of signal transduction
          regulation of phosphatidylinositol 3-kinase signaling
          T cell costimulation
          positive regulation of granulocyte macrophage colony-stimulating factor production
          intracellular signal transduction
          T cell activation
          positive regulation of T-helper 1 cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of alpha-beta T cell proliferation
          viral entry into host cell
          phosphatidylinositol phosphate biosynthetic process
          phosphatidylinositol-mediated signaling
          positive regulation of peptidyl-tyrosine phosphorylation
          cellular response to lipopolysaccharide
          virus receptor activity
          protein binding
          coreceptor activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03030379.hg.1 ENST00000264246.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030379.hg.1 ENST00000478182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030379.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03030379.hg.1 TC03001684.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03030380.hg.1 ENST00000264246.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030380.hg.1 ENST00000478182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030380.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03030380.hg.1 TC03001684.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03030381.hg.1 ENST00000264246.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030381.hg.1 ENST00000478182.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030381.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03030381.hg.1 TC03001684.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03030383.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03030383.hg.1 TC03001684.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03030382.hg.1 ENST00000264246.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03030382.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03030382.hg.1 TC03001684.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190212.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190212.hg.1 TC0300012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300190212.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 TC0300012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190210.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190211.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190211.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190211.hg.1 TC0300012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300190211.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190211.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 TC0300012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190208.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 TC0300012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300190209.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000264246.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000383669.3 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000463729.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 ENST00000478182.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Affymetrix Human Clariom D Assay TC0300012123.hg.1 NM_005191.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 186705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0