Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01046826_m1
          See other MAPK1 GE Assays ›
          Gene Symbol:
          MAPK1
          Gene Name:
          mitogen-activated protein kinase 1
          Gene Aliases:
          ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK
          Chromosome Location:
          Chr.22: 21759657 - 21867680 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01046826_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAPK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002745.4 NP_002736.3 2 - 3 529 72
          NM_138957.3 NP_620407.1 2 - 3 529 72
          GenBank mRNA AK090869.1 - 3 - 4 386 72
          AK295188.1 - 4 - 5 462 72
          AK307615.1 - 3 - 4 374 72
          BC017832.1 - 2 - 3 529 4685557 72
          BC099905.1 - 2 - 3 494 5264558 72
          DQ399292.1 - 2 - 3 289 72
          Z11695.1 - 2 - 3 387 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008105.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000547.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAPK1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          chemotaxis
          response to stress
          cellular response to DNA damage stimulus
          cell cycle
          signal transduction
          chemical synaptic transmission
          axon guidance
          positive regulation of cell population proliferation
          fibroblast growth factor receptor signaling pathway
          response to toxic substance
          positive regulation of peptidyl-threonine phosphorylation
          regulation of phosphatidylinositol 3-kinase signaling
          viral process
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          sensory perception of pain
          cytosine metabolic process
          platelet activation
          positive regulation of cell migration
          thyroid gland development
          regulation of protein stability
          lipopolysaccharide-mediated signaling pathway
          positive regulation of telomere maintenance via telomerase
          regulation of stress-activated MAPK cascade
          mammary gland epithelial cell proliferation
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB signaling pathway
          outer ear morphogenesis
          response to exogenous dsRNA
          response to estrogen
          negative regulation of cell differentiation
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          thymus development
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          regulation of DNA-binding transcription factor activity
          regulation of cytoskeleton organization
          positive regulation of telomerase activity
          Bergmann glial cell differentiation
          long-term synaptic potentiation
          face development
          growth hormone receptor signaling pathway via JAK-STAT
          lung morphogenesis
          trachea formation
          labyrinthine layer blood vessel development
          cardiac neural crest cell development involved in heart development
          ERK1 and ERK2 cascade
          response to epidermal growth factor
          caveolin-mediated endocytosis
          regulation of Golgi inheritance
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          regulation of cellular response to heat
          positive regulation of telomere capping
          regulation of early endosome to late endosome transport
          phosphotyrosine residue binding
          DNA binding
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          transcription factor binding
          RNA polymerase II CTD heptapeptide repeat kinase activity
          kinase activity
          phosphatase binding
          mitogen-activated protein kinase kinase kinase binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 TC22000547.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 ENST00000215832.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 ENST00000398822.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 ENST00000415911.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009889.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000491588.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay TC2200008105.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 NM_138957.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 TC2200008105.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 NM_002745.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000215832.10 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000398822.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 ENST00000544786.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200158004.hg.1 NM_138957.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc