Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022731.hg.1 |
TC19001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022731.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022731.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011052.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192148.hg.1 |
ENST00000222002.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192148.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192148.hg.1 |
TC1900011052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192148.hg.1 |
NM_003167.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192148.hg.1 |
ENST00000222002.3 |
Ensembl |