Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01048278_m1
          See other ELOVL2-AS1 GE Assays ›
          Gene Symbol:
          ELOVL2-AS1
          Gene Name:
          ELOVL2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 11043758 - 11079144 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01048278_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ELOVL2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038962.1 2 - 3 365 92
          NR_038963.1 2 - 3 157 92
          GenBank mRNA BC038757.1 - 2 - 3 365 92
          BC044599.1 - 2 - 3 168 92

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741176 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15785236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15785238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15785237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15785240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741163 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15785233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741171 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04552830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24741112 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 128041 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 139687 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 66409 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 656424 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 376573 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 303694 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 124518 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 176275 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 459367 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 523074 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 4818 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 307894 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 686138 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 731105 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 773041 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 87902 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 262526 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 302039 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143935 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 126362 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 111847 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 AB079384.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 AF202092.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 AK307432.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 AK308919.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 AL137515.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 BC002830.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 BC024221.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR594782.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR601674.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR608289.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR611179.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR619599.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 CR623472.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230574 NM_022488.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 358742 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 256234 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 70644 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 122844 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 68558 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 269408 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 359335 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P557274 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P82204 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 40793 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 35048 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P557274 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P82204 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P17448464 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17448464 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17448501 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17448438 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17448468 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P82204 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P557274 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 32079 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 39340 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 13661 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37516832 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37516810 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37516843 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448501 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15547086 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01422965 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37516859 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448498 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01422986 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37516855 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448438 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15546387 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448464 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448460 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448457 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17448468 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01422942 - -
          Agilent Human 1B Microarray G4111A A_32_P82203 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15547086 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15546387 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17448438 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17448464 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17448468 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17448501 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8116905 - -
          Affymetrix Human Gene 1.1 ST Array 8116904 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030983 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030982 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030982 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030983 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 157335 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline