Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003438.hg.1 |
ENST00000361138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003438.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003438.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003438.hg.1 |
TC09000324.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000475540.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
ENST00000618375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007520.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
NM_015110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
TC0900007520.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
ENST00000361138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
ENST00000471372.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150983.hg.1 |
NM_015110.3 |
Refseq |