Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01048921_m1
          See other RIPK1 GE Assays ›
          Gene Symbol:
          RIPK1
          Gene Name:
          receptor interacting serine/threonine kinase 1
          Gene Aliases:
          RIP, RIP-1, RIP1
          Chromosome Location:
          Chr.6: 3063888 - 3115187 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01048921_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RIPK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005249457.3 XP_005249514.2 1 - 2 97 92
          XM_006715237.3 XP_006715300.2 1 - 2 97 92

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_005249457.3, XM_006715237.3 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          I-kappaB kinase/NF-kappaB signaling
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          positive regulation of necrotic cell death
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          tumor necrosis factor-mediated signaling pathway
          response to tumor necrosis factor
          TRIF-dependent toll-like receptor signaling pathway
          peptidyl-serine autophosphorylation
          positive regulation of apoptotic process
          positive regulation of programmed cell death
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          cellular protein catabolic process
          positive regulation of macrophage differentiation
          positive regulation of transcription by RNA polymerase II
          positive regulation of JNK cascade
          protein autophosphorylation
          positive regulation of NF-kappaB transcription factor activity
          protein homooligomerization
          protein heterooligomerization
          positive regulation of necroptotic process
          T cell apoptotic process
          necroptotic process
          regulation of ATP:ADP antiporter activity
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          death-inducing signaling complex assembly
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          ripoptosome assembly
          necroptotic signaling pathway
          ripoptosome assembly involved in necroptotic process
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of hydrogen peroxide-induced cell death
          amyloid fibril formation
          positive regulation of reactive oxygen species metabolic process
          negative regulation of extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protein kinase activity
          protein serine/threonine kinase activity
          death receptor binding
          protein binding
          ATP binding
          ubiquitin protein ligase binding
          identical protein binding
          protein-containing complex binding
          death domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000259808.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000380409.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 ENST00000479389.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq
          Affymetrix Human Clariom D Assay TC0600006653.hg.1 NM_003804.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0