Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01050628_m1
          See other CEACAM19 GE Assays ›
          Gene Symbol:
          CEACAM19
          Gene Name:
          carcinoembryonic antigen related cell adhesion molecule 19
          Gene Aliases:
          CEACM19, CEAL1
          Chromosome Location:
          Chr.19: 44671452 - 44684355 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01050628_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CEACAM19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127893.1 NP_001121365.1 5 - 6 1187 61
          NM_020219.3 NP_064604.2 5 - 6 1187 61
          GenBank mRNA AK027157.1 - 5 - 6 904 61
          AK128234.1 - 2 - 3 1398 61
          AK291588.1 - 5 - 6 888 61
          AY358394.1 - 5 - 6 917 61
          BC083499.1 - 5 - 6 1187 30390224 61
          KJ851728.1 - 5 - 6 707 61
          KJ851729.1 - 5 - 6 707 61
          KJ851730.1 - 5 - 6 707 61
          KJ851731.1 - 5 - 6 707 61
          KJ851732.1 - 4 - 5 623 61
          KJ851733.1 - 4 - 5 623 61
          KJ851736.1 - 4 - 5 623 61
          KJ851737.1 - 4 - 5 623 61
          KJ851738.1 - 4 - 5 623 61
          KJ851739.1 - 4 - 5 623 61
          KJ851742.1 - 4 - 5 556 61
          KJ851743.1 - 4 - 5 556 61
          KJ851746.1 - 4 - 5 556 61
          KJ851747.1 - 4 - 5 556 61
          KJ851748.1 - 4 - 5 556 61
          KJ851749.1 - 4 - 5 556 61
          KJ851750.1 - 3 - 4 472 61
          KJ851751.1 - 3 - 4 472 61
          KJ851754.1 - 3 - 4 472 61
          KJ851755.1 - 3 - 4 472 61
          KJ851756.1 - 3 - 4 472 61
          KJ851757.1 - 3 - 4 472 61
          KJ851760.1 - 6 - 7 841 61
          KJ851761.1 - 6 - 7 841 61
          KJ851764.1 - 6 - 7 841 61
          KJ851765.1 - 6 - 7 841 61
          KJ851766.1 - 6 - 7 841 61
          KJ851767.1 - 6 - 7 841 61
          KJ851768.1 - 5 - 6 757 61
          KJ851769.1 - 5 - 6 757 61
          KJ851772.1 - 5 - 6 757 61
          KJ851773.1 - 5 - 6 757 61
          KJ851774.1 - 5 - 6 757 61
          KJ851775.1 - 5 - 6 757 61
          KJ851778.1 - 5 - 6 690 61
          KJ851779.1 - 5 - 6 690 61
          KJ851782.1 - 5 - 6 690 61
          KJ851783.1 - 5 - 6 690 61
          KJ851784.1 - 5 - 6 690 61
          KJ851785.1 - 5 - 6 690 61
          KJ851786.1 - 4 - 5 606 61
          KJ851787.1 - 4 - 5 606 61
          KJ851790.1 - 4 - 5 606 61
          KJ851791.1 - 4 - 5 606 61
          KJ851792.1 - 4 - 5 606 61
          KJ851793.1 - 4 - 5 606 61

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CEACAM19

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000358777.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000403660.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000459648.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 ENST00000480278.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_001127893.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011757.hg.1 NM_020219.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline