Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01051882_mH
          See other ITSN1 GE Assays ›
          Gene Symbol:
          ITSN1
          Gene Name:
          intersectin 1
          Gene Aliases:
          ITSN, SH3D1A, SH3P17
          Chromosome Location:
          Chr.21: 33642400 - 33899861 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01051882_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITSN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003024.2 NP_003015.2 36 - 37 4946 123
          XM_005261026.2 XP_005261083.1 35 - 36 5009 123
          XM_017028428.1 XP_016883917.1 37 - 38 4944 123
          XM_017028429.1 XP_016883918.1 36 - 37 4913 123
          XM_017028430.1 XP_016883919.1 35 - 36 4898 123
          XM_017028431.1 XP_016883920.1 35 - 36 4856 123
          XM_017028432.1 XP_016883921.1 34 - 35 4841 123
          XM_017028433.1 XP_016883922.1 34 - 35 4811 123
          XM_017028434.1 XP_016883923.1 33 - 34 4796 123
          XM_017028435.1 XP_016883924.1 34 - 35 4700 123
          XM_017028436.1 XP_016883925.1 33 - 34 4685 123
          XM_017028437.1 XP_016883926.1 31 - 32 4364 123
          GenBank mRNA AF064244.1 - 36 - 37 4784 123
          AF114487.1 - 36 - 37 4946 123
          AY603418.1 - 2 - 3 286 123
          AY605091.1 - 5 - 6 716 123
          BC116185.1 - 35 - 36 4742 40073780 123
          BC116186.2 - 36 - 37 4757 40073781 123
          EU117382.1 - 33 - 34 4497 123
          EU120733.1 - 34 - 35 4664 123
          EU120734.1 - 33 - 34 4496 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000131.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITSN1

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          regulation of Rho protein signal transduction
          positive regulation of phosphorylation
          positive regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          synaptic vesicle endocytosis
          regulation of small GTPase mediated signal transduction
          positive regulation of protein kinase B signaling
          guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          kinase activator activity
          molecular adaptor activity
          proline-rich region binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 TC21000131.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000381285.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000381288.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000381318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000399326.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000399367.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 ENST00000437442.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001102.hg.1 NM_003024.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381284.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381285.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381318.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000399367.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000415023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000420666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 NM_003024.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 TC2100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381285.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381284.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000381318.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000399367.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000415023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 ENST00000420666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146990.hg.1 NM_003024.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0