Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
ENST00000261190.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
ENST00000261191.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
ENST00000335745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
ENST00000539625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017444.hg.1 |
TC12001330.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
TC1200010198.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200177189.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000261191.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000536232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000537336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538155.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000538748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000542392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
ENST00000544548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010198.hg.1 |
NM_018164.2 |
Refseq |