Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01053867_s1
          See other ITPK1-AS1 GE Assays ›
          Gene Symbol:
          ITPK1-AS1
          Gene Name:
          ITPK1 antisense RNA 1
          Gene Aliases:
          C14orf85, ITPK1-AS, ITPK1AS, NCRNA00203
          Chromosome Location:
          Chr.14: 93067452 - 93072152 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01053867_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITPK1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002808.2 1 1495 115
          GenBank mRNA AK024887.1 - 1 1496 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC14000575.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397955 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3891 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3890 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49967 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3397955 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3397955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479076 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940255 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29940265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09479071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479076 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479083 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479068 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479098 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479079 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479074 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479035 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479061 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479103 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16728619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479081 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479107 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479069 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09479071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563524_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234594_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1400008025.hg.1 ENST00000553639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007019.hg.1 ENST00000553639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14007019.hg.1 NR_002808.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14007019.hg.1 TC14000575.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1400008025.hg.1 ENST00000553639.1 Ensembl
          Human Genome U133 Plus 2.0 Array 234594_at - -
          Human Genome U133 Plus 2.0 Array 1563524_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1563524_a_at - -
          Human Genome U133 Plus 2.0 Array 234594_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002808_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563524_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234594_at11 - -
          Human Genome U133 Set 234594_PM_at - -
          Human Genome U133 Set 234594_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051710 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051710 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229818 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2134381 NR_002808.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2134381 AK024887.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2134381 AK024887.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2134381 NR_002808.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2134381 AK024887.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0