Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025825.hg.1 |
ENST00000368466.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025825.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025825.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025825.hg.1 |
TC06002062.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600189849.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189849.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189849.hg.1 |
TC0600013010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189849.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189849.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189851.hg.1 |
TC0600013010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
ENST00000368468.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013010.hg.1 |
NM_005907.3 |
Refseq |