Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01054203_m1
          See other MACF1 GE Assays ›
          Gene Symbol:
          MACF1
          Gene Name:
          microtubule-actin crosslinking factor 1
          Gene Aliases:
          ABP620, ACF7, MACF, OFC4
          Chromosome Location:
          Chr.1: 39084167 - 39487138 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01054203_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MACF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012090.5 NP_036222.3 19 - 20 2423 69
          XM_005270694.1 XP_005270751.1 15 - 16 3084 69
          XM_005270695.1 XP_005270752.1 15 - 16 3084 69
          XM_005270696.1 XP_005270753.1 15 - 16 3084 69
          XM_005270697.1 XP_005270754.1 15 - 16 3084 69
          XM_005270698.1 XP_005270755.1 15 - 16 3084 69
          XM_006710519.1 XP_006710582.1 15 - 16 3084 69
          XM_006710520.2 XP_006710583.1 15 - 16 3084 69
          XM_006710521.1 XP_006710584.1 15 - 16 3084 69
          XM_006710522.1 XP_006710585.1 15 - 16 3084 69
          XM_006710524.2 XP_006710587.1 15 - 16 3084 69
          XM_006710526.3 XP_006710589.1 20 - 21 2516 69
          XM_006710528.3 XP_006710591.1 20 - 21 3077 69
          XM_006710529.3 XP_006710592.1 20 - 21 2434 69
          XM_006710530.3 XP_006710593.1 20 - 21 3322 69
          XM_006710531.3 XP_006710594.1 20 - 21 3053 69
          XM_006710532.3 XP_006710595.1 19 - 20 2981 69
          XM_006710533.2 XP_006710596.1 19 - 20 2981 69
          XM_006710535.3 XP_006710598.1 20 - 21 2586 69
          XM_006710536.2 XP_006710599.1 15 - 16 3084 69
          XM_006710538.2 XP_006710601.1 15 - 16 3084 69
          XM_011541141.2 XP_011539443.1 15 - 16 3084 69
          XM_011541142.2 XP_011539444.1 15 - 16 3084 69
          XM_017000873.1 XP_016856362.1 15 - 16 3084 69
          XM_017000874.1 XP_016856363.1 15 - 16 3084 69
          XM_017000875.1 XP_016856364.1 19 - 20 2406 69
          XM_017000876.1 XP_016856365.1 19 - 20 2490 69
          XM_017000877.1 XP_016856366.1 20 - 21 2412 69
          XM_017000878.1 XP_016856367.1 20 - 21 2275 69
          XM_017000880.1 XP_016856369.1 15 - 16 3084 69
          XM_017000881.1 XP_016856370.1 19 - 20 2398 69
          XM_017000882.1 XP_016856371.1 15 - 16 3084 69
          GenBank mRNA AB029290.1 - 19 - 20 2422 69
          AK127519.1 - 15 - 16 3084 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007898.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000493.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MACF1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of epithelial cell migration
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          regulation of microtubule-based process
          wound healing
          Golgi to plasma membrane protein transport
          positive regulation of axon extension
          regulation of focal adhesion assembly
          cell-cell adhesion
          RNA binding
          actin binding
          calcium ion binding
          protein binding
          microtubule binding
          ATP hydrolysis activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000317713.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000361689.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000372900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000372915.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000404645.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000539005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 ENST00000545844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 NM_012090.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007572.hg.1 TC01000493.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000361689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000372915.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000496804.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000524432.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000530262.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000564288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000567887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 NM_012090.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 TC0100007898.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000361689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000372915.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000496804.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000524432.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000530262.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000564288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 ENST00000567887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155514.hg.1 NM_012090.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0