Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01054219_m1
          See other MACF1 GE Assays ›
          Gene Symbol:
          MACF1
          Gene Name:
          microtubule-actin crosslinking factor 1
          Gene Aliases:
          ABP620, ACF7, MACF, OFC4
          Chromosome Location:
          Chr.1: 39084167 - 39487138 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01054219_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MACF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012090.5 NP_036222.3 44 - 45 6807 101
          XM_005270694.1 XP_005270751.1 45 - 46 13669 101
          XM_005270695.1 XP_005270752.1 45 - 46 13669 101
          XM_005270696.1 XP_005270753.1 45 - 46 13669 101
          XM_005270697.1 XP_005270754.1 45 - 46 13669 101
          XM_005270698.1 XP_005270755.1 45 - 46 13669 101
          XM_006710519.1 XP_006710582.1 45 - 46 13669 101
          XM_006710520.2 XP_006710583.1 45 - 46 13669 101
          XM_006710521.1 XP_006710584.1 45 - 46 13669 101
          XM_006710522.1 XP_006710585.1 45 - 46 13669 101
          XM_006710524.2 XP_006710587.1 44 - 45 13435 101
          XM_006710526.3 XP_006710589.1 50 - 51 13101 101
          XM_006710528.3 XP_006710591.1 50 - 51 13662 101
          XM_006710529.3 XP_006710592.1 50 - 51 13019 101
          XM_006710530.3 XP_006710593.1 50 - 51 13907 101
          XM_006710531.3 XP_006710594.1 50 - 51 13638 101
          XM_006710532.3 XP_006710595.1 49 - 50 13566 101
          XM_006710533.2 XP_006710596.1 49 - 50 13566 101
          XM_006710535.3 XP_006710598.1 50 - 51 13171 101
          XM_006710536.2 XP_006710599.1 40 - 41 7468 101
          XM_006710538.2 XP_006710601.1 45 - 46 13669 101
          XM_011541141.2 XP_011539443.1 45 - 46 13615 101
          XM_011541142.2 XP_011539444.1 44 - 45 8218 101
          XM_017000873.1 XP_016856362.1 45 - 46 13669 101
          XM_017000874.1 XP_016856363.1 45 - 46 13669 101
          XM_017000875.1 XP_016856364.1 49 - 50 12991 101
          XM_017000876.1 XP_016856365.1 49 - 50 13075 101
          XM_017000877.1 XP_016856366.1 50 - 51 12997 101
          XM_017000878.1 XP_016856367.1 50 - 51 12860 101
          XM_017000879.1 XP_016856368.1 15 - 16 8431 101
          XM_017000880.1 XP_016856369.1 40 - 41 7468 101
          XM_017000881.1 XP_016856370.1 44 - 45 6782 101
          XM_017000882.1 XP_016856371.1 40 - 41 7468 101
          GenBank mRNA AB007934.3 - 24 - 25 4155 101
          AB029290.1 - 44 - 45 6806 101
          AB033077.1 - 24 - 25 4155 101
          AF317696.1 - 14 - 15 8381 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007898.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000493.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MACF1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of epithelial cell migration
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          regulation of microtubule-based process
          wound healing
          Golgi to plasma membrane protein transport
          positive regulation of axon extension
          regulation of focal adhesion assembly
          cell-cell adhesion
          RNA binding
          actin binding
          calcium ion binding
          protein binding
          microtubule binding
          ATP hydrolysis activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000289893.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000317713.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000361689.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000372900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000372915.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000476350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000539005.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 ENST00000545844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 NM_012090.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007612.hg.1 TC01000493.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000289893.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000361689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000372915.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000372925.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000476350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000564288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000567887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 NM_012090.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 TC0100007898.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000289893.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000361689.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000372915.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000372925.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000476350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000564288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 ENST00000567887.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155557.hg.1 NM_012090.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0