Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01054248_m1
See other MACF1 GE Assays ›
Gene Symbol:
MACF1
Gene Name:
microtubule-actin crosslinking factor 1
Gene Aliases:
ABP620, ACF7, MACF, OFC4
Chromosome Location:
Chr.1: 39084167 - 39487138 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01054248_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MACF1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005270694.1
XP_005270751.1
91 - 92
22507
74
XM_005270696.1
XP_005270753.1
91 - 92
22507
74
XM_005270698.1
XP_005270755.1
91 - 92
22507
74
XM_006710519.1
XP_006710582.1
90 - 91
22498
74
XM_006710520.2
XP_006710583.1
90 - 91
22444
74
XM_006710521.1
XP_006710584.1
91 - 92
22507
74
XM_006710524.2
XP_006710587.1
90 - 91
22273
74
XM_006710526.3
XP_006710589.1
96 - 97
21939
74
XM_006710528.3
XP_006710591.1
96 - 97
22500
74
XM_006710529.3
XP_006710592.1
96 - 97
21857
74
XM_006710530.3
XP_006710593.1
96 - 97
22745
74
XM_006710531.3
XP_006710594.1
96 - 97
22476
74
XM_006710532.3
XP_006710595.1
95 - 96
22404
74
XM_006710533.2
XP_006710596.1
95 - 96
22404
74
XM_006710535.3
XP_006710598.1
96 - 97
22009
74
XM_006710536.2
XP_006710599.1
86 - 87
16306
74
XM_011541141.2
XP_011539443.1
91 - 92
22453
74
XM_011541142.2
XP_011539444.1
90 - 91
17056
74
XM_017000873.1
XP_016856362.1
90 - 91
22498
74
XM_017000875.1
XP_016856364.1
95 - 96
21829
74
XM_017000876.1
XP_016856365.1
95 - 96
21913
74
XM_017000877.1
XP_016856366.1
96 - 97
21835
74
XM_017000878.1
XP_016856367.1
96 - 97
21698
74
XM_017000879.1
XP_016856368.1
61 - 62
17269
74
XM_017000880.1
XP_016856369.1
85 - 86
16243
74
XM_017000881.1
XP_016856370.1
90 - 91
15620
74
GenBank mRNA
AF317696.1
-
60 - 61
17219
74
AK297854.1
-
3 - 4
180
74
BC071925.1
-
4 - 5
298
74
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297854.1 transcripts.
Set Membership:
Amplicon length between 71 and 85
3' Most
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
regulation of epithelial cell migration
Wnt signaling pathway
positive regulation of Wnt signaling pathway
regulation of microtubule-based process
wound healing
Golgi to plasma membrane protein transport
positive regulation of axon extension
regulation of focal adhesion assembly
cell-cell adhesion
RNA binding
actin binding
calcium ion binding
protein binding
microtubule binding
ATP hydrolysis activity
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0