Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002133.hg.1 |
ENST00000378357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002133.hg.1 |
ENST00000544074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002133.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002133.hg.1 |
TC09000191.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
TC0900007094.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148207.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000378357.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000485665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000493245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
ENST00000617161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007094.hg.1 |
NM_001216.2 |
Refseq |