Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000342198.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000347433.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000388793.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000432932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000457580.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
ENST00000539682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
NM_001128852.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
NM_001128853.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
NM_001128854.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
NM_015908.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009916.hg.1 |
TC07000636.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000445337.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000448764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000477529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000478693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000611405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000614484.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000618262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000618411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128853.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_015908.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
TC0700008568.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000478693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000445337.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000448764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000477529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000611405.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000614484.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000618262.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
ENST00000618411.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128852.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128853.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_001128854.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160579.hg.1 |
NM_015908.5 |
Refseq |