Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01056533_m1
          See other IRF4 GE Assays ›
          Gene Symbol:
          IRF4
          Gene Name:
          interferon regulatory factor 4
          Gene Aliases:
          LSIRF, MUM1, NF-EM5, SHEP8
          Chromosome Location:
          Chr.6: 391739 - 411443 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01056533_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IRF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195286.1 NP_001182215.1 5 - 6 761 60
          NM_002460.3 NP_002451.2 5 - 6 764 60
          NR_046000.2 5 - 6 761 60
          GenBank mRNA AK130762.1 - 2 - 3 655 60
          AK310149.1 - 5 - 6 751 60
          BC015752.1 - 5 - 6 711 4861223 60
          D78261.1 - 5 - 6 730 60
          U52682.1 - 5 - 6 760 60
          U63738.1 - 5 - 6 743 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006442.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000005.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IRF4

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          peptidyl-lysine methylation
          negative regulation of toll-like receptor signaling pathway
          T cell activation
          defense response to protozoan
          myeloid dendritic cell differentiation
          positive regulation of DNA binding
          histone H3 acetylation
          histone H4 acetylation
          positive regulation of interleukin-10 biosynthetic process
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of interleukin-13 biosynthetic process
          positive regulation of interleukin-4 biosynthetic process
          regulation of T-helper cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          T-helper 17 cell lineage commitment
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein-lysine N-methyltransferase activity
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000380956.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000468485.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000469834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000493114.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 ENST00000495137.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_001195286.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NM_002460.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006442.hg.1 NR_046000.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0