Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015827.hg.1 |
ENST00000264167.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015827.hg.1 |
ENST00000536686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015827.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015827.hg.1 |
TC02001065.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170753.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170753.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170753.hg.1 |
TC0200010100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200170753.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170753.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000264167.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000409888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
ENST00000460342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010100.hg.1 |
NM_003659.3 |
Refseq |