Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003728.hg.1 |
TC20000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003728.hg.1 |
ENST00000279022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003728.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003728.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003728.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003730.hg.1 |
TC20000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003730.hg.1 |
ENST00000279022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003730.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003730.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003730.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
TC2000007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150087.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
TC2000007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150084.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007251.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
NM_181526.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
TC2000007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
ENST00000279022.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
ENST00000346786.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
NM_006097.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150086.hg.1 |
NM_181526.2 |
Refseq |