Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01058356_m1
          See other DGUOK GE Assays ›
          Gene Symbol:
          DGUOK
          Gene Name:
          deoxyguanosine kinase
          Gene Aliases:
          MTDPS3, dGK
          Chromosome Location:
          Chr.2: 73926826 - 73958961 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01058356_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DGUOK
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318860.1 NP_001305789.1 1 - 2 233 76
          NM_001318863.1 NP_001305792.1 1 - 2 233 76
          NR_134893.1 1 - 2 233 76
          NR_134894.1 1 - 2 233 76
          NR_134896.1 1 - 2 233 76
          NR_134898.1 1 - 2 233 76
          GenBank mRNA BC001121.2 - 1 - 2 172 76
          BC024019.1 - 1 - 2 187 76
          BF975904.1 - 1 - 2 178 76
          BG708680.1 - 1 - 2 222 76
          BG715600.1 - 1 - 2 174 76
          BM706344.1 - 1 - 2 164 76
          KU178024.1 - 1 - 2 148 76

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC001121.2, NM_001318860.1, NM_001318863.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          guanosine metabolic process
          deoxyribonucleoside monophosphate biosynthetic process
          nucleotide biosynthetic process
          negative regulation of neuron projection development
          purine-containing compound salvage
          dGTP metabolic process
          purine deoxyribonucleoside metabolic process
          deoxyguanosine kinase activity
          ATP binding
          nucleoside kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000264093.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000348222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000418996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462551.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000462685.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000489796.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000493055.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 ENST00000629438.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080916.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Affymetrix Human Clariom D Assay TC0200008063.hg.1 NM_080918.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 217726 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0